Check Order Status
BBL™ Gram Slide
Actual product may vary
Fisherbrand
BBL™ Gram Slide
Used for evaluating and controlling the quality of Gram stain reagents and techniques. BD Difco™ BBL™ Gram Slide enables easy evaluations.
Have Questions?
Catalog Number
Quantity
FISB231401
50 Pk.
Catalog number
FISB231401
Price (SGD)
-
Quantity:
50 Pk.
Slide format
Used for evaluating: Gram stain reagents
50 slides
Specifications
Quantity
50 Pk.
Unit Size
PK
Contents & Storage
30°C