BBL™ IsoVitaleX™ Enrichment
BBL™ IsoVitaleX™ Enrichment
Actual product may vary
Fisherbrand
BBL™ IsoVitaleX™ Enrichment
BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.
Have Questions?
Catalog NumberQuantity
FISB1187610 mL
Catalog number FISB11876
Price (SGD)
-
Quantity:
10 mL
  • GC Agar supplemented with hemoglobin and BBL IsoVitaleX Enrichment (in place of yeast concentrate) provides an improved medium for the isolation of microorganisms
  • Enrichment provides V factor (nicotinamide adenine dinucleotide, NAD) for Haemophilus species
  • Also provides vitamins, amino acids, coenzymes, dextrose, ferric ions and other factors which improve the growth of pathogenic Neisseria
Specifications
Certifications/ComplianceISO/DIS, 11014
AdditiveIsoVitaleX™ Enrichment
FormLyophilized
For Use With (Application)Dehydrated culture media
Packaging TypeVial
Quantity10 mL
Shelf Life1095 Days
Unit SizePK
Contents & Storage
2°C to 8°C