▸
Histology and Cytology
▸
Tissue Embedding
▸
Embedding Cassettes and Storage
▸
Modular Cassette Storage Drawers
Actual product may vary
Thermo Scientific
Modular Cassette Storage Drawers
Permanently store up to 165 embedding rings or 250 cassettes per drawer with modular cassette storage drawers.
Have Questions?
Catalog Number
Product Type
FISB1001304
Cassette Storage Drawer
Catalog number
FISB1001304
Price (SGD)
-
Product Type:
Cassette Storage Drawer
Features
Made from high-impact polystyrene
Designed with Interlocking ridges on top and bottom to permit stacking to any convenient height
Includes
Identification labels.
Specifications
Product Type
Cassette Storage Drawer
Holds
250