Drosophila Bottles
Drosophila Bottles
Actual product may vary
Drosophila Bottles
Wide base/narrow top design prevents fly meal from falling out when bottle is turned upside down
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeCapacity (English)Material
FISAS359Square6 oz.Polyethylene
FISAS117Round8 oz.Polypropylene
Catalog number FISAS359
Price (SGD)
666.00
CS
Add to cart
Bottom Shape:
Square
Capacity (English):
6 oz.
Material:
Polyethylene
Price (SGD)
666.00
CS
Add to cart
Wide base/narrow top design prevents fly meal from falling out when bottle is turned upside down
Drosophila bottles are available in transparent, autoclavable polypropylene and non-autoclavable polyethylene. The bottle openings are designed to accept standard paper lids, foam plugs, cotton balls and Buzz Plugs.
Round-Bottom Bottles
  • Polypropylene
  • Single use or autoclave for reuse
Square-Bottom Bottles
  • Available in polypropylene or polyethylene
  • Polypropylene provides good transparency and can be autoclaved for reuse or disposed of after single use
Specifications
Bottom ShapeSquare
Capacity (English)6 oz.
Capacity (Metric)177 mL
ColorTransparent
For Use With (Application)Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Height (English)4 in.
Height (Metric)103 mm
Length (English)2.24 in.
Length (Metric)57 mm
Width (English)2.24 in.
Width (Metric)57 mm
MaterialPolyethylene
TypeStock Bottle
Unit SizeCS