Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Actual product may vary
Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
FISAS2752.8cm wide plastic shell vials
FISAS2732.5cm narrow plastic shell vials
FISAS271Standard (2.4cm dia.) glass vials
Catalog number FISAS275
Price (SGD)
329.00
CS
Add to cart
For Use With (Equipment):
2.8cm wide plastic shell vials
Price (SGD)
329.00
CS
Add to cart
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
For Use With (Application)Minimizes evaporation, keeps food fresh longer
For Use With (Equipment)2.8cm wide plastic shell vials
IncludesSuperior barrier
Quantity1000 Case
MaterialCellulose Acetate
TypeBuzzPlugs Bottle Closure
Unit SizeCS