Trays and Dividers for Shell Vials
Trays and Dividers for Shell Vials
Actual product may vary
Trays and Dividers for Shell Vials
Accommodate narrow- or wide-diameter vials.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)
FISAS260Tray for Narrow VialsFor narrow vials (25 x 95 mm)
Catalog number FISAS260
Price (SGD)
361.00
CS
Add to cart
Type:
Tray for Narrow Vials
For Use With (Equipment):
For narrow vials (25 x 95 mm)
Price (SGD)
361.00
CS
Add to cart
  • Cardboard trays have cell dividers
  • 10 x 10 in. (25 x 25cm) row configuration
Specifications
AutoclavableNot Autoclavable
For Use With (Equipment)For narrow vials (25 x 95 mm)
IncludesDividers
MaterialCardboard
TypeTray for Narrow Vials
Unit SizeCS