Drosophila Products Jazz-Mix™ Drosophila Food
Drosophila Products Jazz-Mix™ Drosophila Food
Actual product may vary
Drosophila Products Jazz-Mix™ Drosophila Food
Simply add water—conveniently packaged as a complete and quick-to-prepare mixture.
Have Questions?
Catalog number FISAS153
Price (SGD)
277.00
EA
Add to cart
Price (SGD)
277.00
EA
Add to cart
  • Consists of brown sugar, corn meal, yeast, agar, benzoic acid, methyl paraben and propionic acid
Notes: Product is not made for human consumption. Yield may vary based on evaporation while boiling.
Specifications
Weight (English)10 lb.
Weight (Metric)4.5 kg
TypeJazzmix Drosophila Food
Unit SizeEA