Glass Fiber Filters Without Binder, 1.2 μm
Glass Fiber Filters Without Binder, 1.2 μm
Actual product may vary
MilliporeSigma
Glass Fiber Filters Without Binder, 1.2 μm
Hydrophilic Glass Fiber without Binder Resin filter
Have Questions?
Catalog NumberDiameter (Metric)Grade
FISAPFC047004.7 cmAPFC
Catalog number FISAPFC04700
Price (SGD)
301.00
PK
Add to cart
Diameter (Metric):
4.7 cm
Grade:
APFC
Price (SGD)
301.00
PK
Add to cart
Hydrophilic Glass Fiber without Binder Resin filter
Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).
Glass Fiber Filters without Binder Resin:
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
Type APFA
•Retains fine particles with good efficiency, even at high flow rates
•Recommended for monitoring wastewater and collecting suspended particles in gases (e.g., fumes from chimneys)
•Use in biochemical applications (e.g., collection of cells and filtration of protein or nucleic acid precipitates)
Specifications
Airflow Rate41.2 L/min/ cm2
Air Resistance54 mm of H2O
AutoclavableAutoclavable
ColorWhite
Filter TypeDepth Filter
MaterialBorosilicate Glass Microfiber
Particle Retention1.2 μm
Pore Size1.2 μm
SterilitySterile
Flow Rate1.2 mL/min/ cm2
For Use With (Application)Determination of total suspended solids in drinking water, Filtration of proteins of nucleic acid TCA precipitates
GradeAPFC
HydrophobicityHydrophilic
Surface TypePlain
Temperature (English) Max.932°F
Temperature (Metric) Max500°C
Thickness (English)0.0094 in.
Thickness (Metric)240 μm
TypeFilter Membranes
WettabilityHydrophilic
Diameter (English)1.8 in.
Diameter (Metric)4.7 cm
Diameter (Metric) Disc4.7 cm
Unit SizePK