Glass Fiber Filters with Binder, 1 μm
Glass Fiber Filters with Binder, 1 μm
Actual product may vary
MilliporeSigma
Glass Fiber Filters with Binder, 1 μm
For prefiltration and contamination analysis
Have Questions?
Catalog NumberDiameter (Metric)
FISAP15047004.7 cm
Catalog number FISAP1504700
Price (SGD)
273.00
PK
Add to cart
Diameter (Metric):
4.7 cm
Price (SGD)
273.00
PK
Add to cart
For prefiltration and contamination analysis
Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121°C at 1 bar). Glass Fiber Filters with Binder Resin: Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.
Type AP15
  • Lowest dirt-holding capacity
  • Recommended prefilter for 0.2 to 0.6 μm filters
  • Specifications
    Airflow Rate10.6 L/min/ cm2
    Air Resistance210 mm of H2O
    AutoclavableAutoclavable
    ColorWhite
    Filter TypeDepth Filter
    MaterialBorosilicate Glass Microfiber
    Particle Retention0.2 to 0.6 μm
    Pore Size1 μm
    SterilitySterile
    Flow Rate1.6 mL/min/ cm2
    For Use With (Application)Prefiltration and contamination analysis
    GradeAP15
    HydrophobicityHydrophilic
    IncludesBinder resin
    Protein Binding Capacity100 μg/ cm2
    Surface TypePlain
    Temperature (English) Max.1022°F
    Temperature (Metric) Max550°C
    Thickness (English)0.031 in.
    Thickness (Metric)790 μm
    TypeFilter Membranes
    WettabilityHydrophilic
    Diameter (English)1.8 in.
    Diameter (Metric)4.7 cm
    Diameter (Metric) Disc4.7 cm
    Unit SizePK