Potassium AA Standard, 1000 ppm K in H2O, Ricca Chemical
Potassium AA Standard, 1000 ppm K in H<sub>2</sub>O, Ricca Chemical
Potassium AA Standard, 1000 ppm K in H<sub>2</sub>O, Ricca Chemical
Potassium AA Standard, 1000 ppm K in H<sub>2</sub>O, Ricca Chemical
Potassium AA Standard, 1000 ppm K in H<sub>2</sub>O, Ricca Chemical
Potassium AA Standard, 1000 ppm K in H2O, Ricca Chemical
Have Questions?
Catalog NumberQuantityPackaging
FISAK1KW500500 mLNatural Poly Bottle
Catalog number FISAK1KW500
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium AA Standard
Shelf Life24 months
PackagingNatural Poly Bottle
CAS7447-40-7
CAS Min %98.0
View more