Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
Buffer solution pH 4, Citrate buffer
CAS: 68-04-2 | C8H5KO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number FISAC258580010
Price (SGD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS68-04-2, 7647-14-5, 7732-18-5
Specifications
Chemical Name or MaterialBuffer solution pH 4
Name NotePhthalate buffer
CAS68-04-2
ColorColorless
CAS7647-14-5
View more
pH buffer
RUO – Research Use Only