Easy Pierce Heat Sealing Foil
Easy Pierce Heat Sealing Foil
Actual product may vary
Thermo Scientific
Easy Pierce Heat Sealing Foil
Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
FISAB1720AluminumEasyPierce 20μm Thin Foil85 x 135 mm
AB-0757AluminumEasy Pierce Foil85 x 135 mm
FISAB-0757AluminumEasy Pierce Foil85 x 135 mm
FISAB-0559AluminumThermo-Seal85 x 135 mm
Catalog number FISAB1720
Price (SGD)
492.00
PK
Add to cart
Material:
Aluminum
Product Type:
EasyPierce 20μm Thin Foil
Dimensions (L x W):
85 x 135 mm
Price (SGD)
492.00
PK
Add to cart
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20μm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
Specifications
ColorFoil
Solvent Or Alcohol ResistantLife Science Reagents Resistant, DMSO Resistant
MaterialAluminum
SterilityNon-sterile
AutoclavableNot Autoclavable
No. of Wells96
Dimensions (L x W)85 x 135 mm
For Use With (Application)Ideal for compound management and high throughput screening applications, suitable for PCR
Product TypeEasyPierce 20μm Thin Foil
Product LineAbgene
AdhesivePolypropylene
Certified RNase/DNase-freeNo
PierceableYes
Temperature Range (Metric)-80°C to +80°C, -80°C to +120°C (with Adequate Pressure)
Well ShapeAll well shapes
Plate CompatibilityPolypropylene, Polyethylene
Incubation ConditionsGeneral
PeelableYes
Unit SizePK
Contents & Storage
General; Long term