Abgene™ SmartCap Mats
Abgene™ SmartCap Mats
Actual product may vary
Abgene™ SmartCap Mats
Abgene™ SmartCap Mats
Abgene™ SmartCap Mats
Abgene™ SmartCap Mats
Abgene™ SmartCap Mats
The Thermo Scientific™ Abgene™ SmartCap Mat ensures reliable sealing from ambient temperatures down to -80 °C.
Have Questions?
Catalog NumberDescription
FISAB1413Seals 96 sample tubes at one time
Catalog number FISAB1413
Price (SGD)
660.00
CS
Add to cart
Description:
Seals 96 sample tubes at one time
Price (SGD)
660.00
CS
Add to cart
The Thermo Scientific™ Abgene™ SmartCap Mat seals 96 samples at one time in a high-density storage format,affording maximum space saving benefits. Suitable for use with Thermo Scientific storage tubes ranging from 0.3 ml to1.4 ml, the Abgene SmartCap Mat maintain sa consistent seal down to -80°C.
Features:
  • Efficient Storage
  • Enhanced Materials
  • Ease of Installation and Access
Specifications
DescriptionSeals 96 sample tubes at one time
No. of Wells96
Product TypeSealing Mat
Temperature Range (Metric)-80°C
Unit SizeCS