PCR Plate, 96-well, segmented, semi-skirted, yellow
PCR Plate, 96-well, segmented, semi-skirted, yellow
Actual product may vary
Fisherbrand
PCR Plate, 96-well, segmented, semi-skirted, yellow
Patented segmented design allows plate to be cut into 24- and 48-well sections
Have Questions?
Catalog NumberQuantity
FISAB0900Y25 Plates
Catalog number FISAB0900Y
Price (SGD)
323.00
CS
Add to cart
Quantity:
25 Plates
Price (SGD)
323.00
CS
Add to cart
  • Virgin polypropylene
  • Raised rim, thin-wall design
  • Clear well bottom for sample visibility
  • Compatible with all major thermal cyclers
  • 300μL maximum well volume (when used with adhesive and heat seals)
  • Cut-away corner and alphanumeric grid
  • Available in a rigid version for robotic handling applications; rigid version cannot be cut
  • Patented segmented plate design allows plates to be cut into 24- and 48-well sections
  • Semi-skirt adds rigidity and allows for labeling or bar coding
  • Cut corner: H1
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase/DNase-free
Packaging25 Plate
ColorYellow
Product TypePCR Microplate
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Skirt StyleSemi
MaterialPolypropylene
Volume (Metric) Well0.3 mL
No. of Wells96
Volume (Metric) Working25 μL
Quantity25 Plates
Reaction SpeedStandard
Unit SizeCS