Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Actual product may vary
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
Thermo Scientific
Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
384-well microarray plate with pyramidal well bottoms for maximum sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WellWell ShapeFit for Purpose
FISAB-1178Natural250 μLSquare, Conical Bottom (V-well)PCR Ready
AB-0781Natural120 μLSquare, Conical Bottom (V-well)PCR Ready
AB-1178Natural250 μLSquare, Conical Bottom (V-well)PCR Ready
Catalog number FISAB-1178
Price (SGD)
1,033.00
CS
Add to cart
Color:
Natural
Volume (Metric) Well:
250 μL
Well Shape:
Square, Conical Bottom (V-well)
Fit for Purpose:
PCR Ready
Price (SGD)
1,033.00
CS
Add to cart
Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.
Highlights
  • Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
  • Pyramidal well bottom for ease of sample retrieval
  • Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination
Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported
All plates are ANSI format for compatibility with automated systems.
To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
FormatStandard
Description384-well Polypropylene Sample Processing & Storage Plates
Array16 x 24
AutoclavableAutoclavable
Packaging10 pk., 50 cs.
Binding TypeLow binding PP
Product LineAbgene
ColorNatural
Product Type384-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well250 μL
No. of Wells384
Volume (Metric) Working200 μL
SterilityCleanroom
Surface TreatmentNon-treated
Well ShapeSquare, Conical Bottom (V-well)
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF5000 x g
No. per Case50
No. per Pack10
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
RNase FreeYes
Fit for PurposePCR Ready
Plate SizeANSI/ SLAS Standard
Unit SizeCS