Domed PCR Caps, strips of 12
Domed PCR Caps, strips of 12
Actual product may vary
Fisherbrand
Domed PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Have Questions?
Catalog NumberQuantityColor
FISAB-085325 packs of 8Clear
Catalog number FISAB-0853
Price (SGD)
336.00
CS
Add to cart
Quantity:
25 packs of 8
Color:
Clear
Price (SGD)
336.00
CS
Add to cart
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
ColorClear
MaterialPolypropylene
No. of Wells12
Quantity25 packs of 8
FormatStrips of 12 Domed Caps
Product TypePCR Plate Cap Strip
Certified RNase/DNase-freeYes
Plate Compatibility96-well Plate
Unit SizeCS