Flat PCR Cap Strips Promotion
Product Image
Fisherbrand
Flat PCR Cap Strips Promotion
Optimize PCR applications with these flat cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Have Questions?
Catalog NumberQuantity
FISAB-0784250/Cs.
Catalog number FISAB-0784
Price (SGD)
229.00
CS
Add to cart
Quantity:
250/Cs.
Price (SGD)
229.00
CS
Add to cart
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
0
Specifications
ColorClear Frosted
Certifications/ComplianceCertified free of human DNA, RNase, DNase, and endotoxins.
MaterialVirgin Medical Grade Polypropylene
No. of Wells96
No. per Case250
No. per Pack250
Quantity250/Cs.
For Use With (Application)Facilitates sample labeling
FormatStrips of 8 Flat Caps
Plate Compatibility96-well PCR Plate, Standard and Low-profile PCR Tube Strips
Unit SizeCS