Thermo Scientific™ Abgene™ Blank and Alphanumeric Storage Tubes
Product Image
Thermo Scientific
Thermo Scientific™ Abgene™ Blank and Alphanumeric Storage Tubes
Thermo Scientific™ ABgene™ Blank and Alphanumeric Storage Tubes enable secure sample storage, while allowing individual access, sealing and retrieval of samples.
Have Questions?
Catalog NumberVolume (Metric)Packaging
FISAB-06721.2 mLBulk, 1000 tubes/cs
Catalog number FISAB-0672
Price (SGD)
425.00
CS
Add to cart
Volume (Metric):
1.2 mL
Packaging:
Bulk, 1000 tubes/cs
Price (SGD)
425.00
CS
Add to cart
Thermo Scientific™ ABgene™ Blank and Alphanumeric Storage Tubes enable secure sample storage, while allowing individual access, sealing and retrieval of samples. It includes Abgene™ 1.2mL open top storage tube in bulk pack (Cat # AB-0672) and Abgene™ 0.65mL open top storage tubes in racks (Cat # AB-1256).
Features
  • Can be picked and placed for reformatting of sample selections and eliminate the need to thaw an entire storage plate for only a few desired samples
  • Manufactured from 100% virgin, medical-grade polypropylene and every lot is leak tested to ensure the integrity and security of samples
  • Available blank or with alphanumeric markings molded into the bottom of each tube for enhanced sample identification
  • Wide variety of tube volumes and capping options available, both in bulk or racked V-well design for improved sample recovery
  • Open bottom rack design for faster freezing and thawing
  • Broad temperature range of -180° to 120°C
Enhanced Storage Rack Design
  • Unlike traditional storage tubes or blocks, blank and alphanumeric storage tubes are available in specially designed, stackable, microplate-footprint racks of 96 tubes for individual sample, access and removal
  • Rack design includes grid-referenced lids for easy identification of tube location
Specifications
Volume (Metric)1.2 mL
SterilityNon-sterile
FormatBulk
Tip StyleUniversal Fit
PackagingBulk, 1000 tubes/cs
Product LineAbgene