Tubes, strips of 8
Tubes, strips of 8
Actual product may vary
Fisherbrand
Tubes, strips of 8
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
Have Questions?
Catalog NumberColorClosure TypeProduct TypeQuantity
FISAB-0264NaturalNot Included8-Strip Tube250 Strips
Catalog number FISAB-0264
Price (SGD)
583.00
CS
Add to cart
Color:
Natural
Closure Type:
Not Included
Product Type:
8-Strip Tube
Quantity:
250 Strips
Price (SGD)
583.00
CS
Add to cart
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Specifications
Certifications/ComplianceCertified DNA/RNase Free
ColorNatural
Volume (Metric) Working0.2 mL
Volume (Metric)0.25 mL
Quantity250 Strips
For Use With (Equipment)Thermal Cyclers
MaterialPolypropylene
Reaction SpeedStandard
Closure TypeNot Included
Format250 Strip Tubes
Product Type8-Strip Tube
Unit SizeCS