Phenol (Crystals), Fisher Chemical™
Phenol (Crystals), Fisher Chemical™
Phenol (Crystals), Fisher Chemical™
Phenol (Crystals), Fisher Chemical™
Phenol (Crystals), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FISA92-100100 gAmber Glass/Poison Pack
Catalog number FISA92-100
Price (SGD)
-
Quantity:
100 g
Packaging:
Amber Glass/Poison Pack
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenol
CAS108-95-2
ChemAlert Storage SymbolWhite
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:3
Flammability:2
Instability:1
View more