Urine Test Strips, Medi‐Test Combi 5
Urine Test Strips, Medi‐Test Combi 5
Actual product may vary
Urine Test Strips, Medi‐Test Combi 5
Urine test strips, Medi‐Test Combi 5, pack of 100 strips
Have Questions?
Catalog NumberNo. per Pack
FIS930-32100 Pk.
Catalog number FIS930-32
Price (SGD)
-
No. per Pack:
100 Pk.
The use of urine test strips is acknowledged as modern screening method in medical practice. Within minutes important information on the health status of the patient is obtained. This simplifies the decision about further diagnostic and therapeutic action.
  • Parameters: Ascorbic acid, Blood, Glucose, pH, Protein
  • Ascorbic acid: neg, +, ++
  • Blood: neg, ca. 5–10, ca. 50, ca. 250
  • Glucose: neg, norm, 50, 150, 500, >1000
  • pH: 5 to 9
  • Protein: neg, 30, 100, 500
Specifications
No. per Pack100 Pk.
PackagingBox
Unit SizeEA