Check Order Status
Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical
Element Search
Structure Search
Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS9150-1
4 L
Natural Poly Bottle
Catalog number
FIS9150-1
Price (SGD)
-
Quantity:
4 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Water
Shelf Life
18 months
Name Note
ASTM Type I
CAS
7732-18-5
Color
Colorless
View more