▸
pH and Electrochemistry
▸
pH Paper and other Test Strips
▸
pH Test Strips, Ricca Chemical
Actual product may vary
Ricca Chemical
pH Test Strips, Ricca Chemical
General-purpose pH test strips
Have Questions?
Catalog Number
pH Range
Graduations
Quantity
FIS88-841
2 to 9
0.5
1 Ea.
Catalog number
FIS88-841
Price (SGD)
-
pH Range:
2 to 9
Graduations:
0.5
Quantity:
1 Ea.
Available in four different pH ranges
Time-saving, single-use indicator strips with color-fixed test area on an easy-to-handle, durable plastic strip
Fast accurate results
Packages include a color comparison chart
100 strips per pack
Specifications
pH Range
2 to 9
For Use With (Application)
For precise pH determination
Graduations
0.5
Includes
Color chart
Quantity
1 Ea.
Shelf Life
4 Years
Storage Requirements
15°C to 30°C
Type
pH Test Strip