Check Order Status
Total Suspended Solids (TSS) Standard, 100 ppm TSS (Gravimetric, Dried at 103-105°C), Ricca Chemical
Element Search
Structure Search
Total Suspended Solids (TSS) Standard, 100 ppm TSS (Gravimetric, Dried at 103-105°C), Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS8672-16
500 mL
Catalog number
FIS8672-16
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Total Suspended Solids (TSS) Standard
Packaging
Amber Glass Bottle
CAS
9004-34-6
CAS Min %
98.0
Concentration or Composition Notes
100mg/L (100ppm) TSS
View more