Check Order Status
Synthetic Seawater, ASTM D 1141 Substitute Ocean Water without Heavy Metals, Ricca Chemical
Element Search
Structure Search
Synthetic Seawater, ASTM D 1141 Substitute Ocean Water without Heavy Metals, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS8363-1
4 L
Catalog number
FIS8363-1
Price (SGD)
-
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or Material
Synthetic Sea Water
CAS
7732-18-5
CAS Min %
94.0
CAS Max %
98.0
CAS
7647-14-5
View more