Check Order Status
Starch Indicator, 1% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Element Search
Structure Search
Starch Indicator, 1% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Iodometric Titrations
Have Questions?
Change view
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
Catalog number
FIS8050-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7732-18-5, 9005-84-9, 69-72-7
Specifications
Chemical Name or Material
Starch Indicator
CAS
7732-18-5
CAS Min %
97.0
CAS Max %
100.0
CAS
9005-84-9
View more