Check Order Status
Sodium Hydroxide, 0.100 N (N/10), Ricca Chemical
Element Search
Structure Search
Sodium Hydroxide, 0.100 N (N/10), Ricca Chemical
Standardized Bases
Have Questions?
Catalog Number
Quantity
Packaging
FIS7350-32
1 L
Natural Poly Bottle
Catalog number
FIS7350-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
CAS
1310-73-2
CAS Min %
97%
CAS Max %
100%
CAS
7732-18-5
View more
Tested in an ISO 17025 Accredited Facility
Test Reports provided with Certified Values and Measurement Uncertainty
Tight release specifications for low lot-to-lot variability