Check Order Status
Sodium Azide, 5% (w/v), Ricca Chemical
Element Search
Structure Search
Sodium Azide, 5% (w/v), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS71448-16
500 mL
Natural Poly Bottle
Catalog number
FIS71448-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Azide
CAS
26628-22-8
CAS Min %
93.0
CAS Max %
97.0
CAS
7732-18-5
View more