Check Order Status
Ammonium Oxalate TS, 3.5% (w/v), Ricca Chemical
Element Search
Structure Search
Ammonium Oxalate TS, 3.5% (w/v), Ricca Chemical
Meets USP TS and ACS RS specifications
Have Questions?
Catalog Number
Quantity
Packaging
FIS679-16
500 mL
Natural Poly Bottle
Catalog number
FIS679-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
AMMONIUM OXALATE
CAS
6009-70-7
CAS Min %
95.0
CAS Max %
99.0
CAS
7732-18-5
View more