▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Shandon™ Bluing Reagent
Actual product may vary
Thermo Scientific
Shandon™ Bluing Reagent
Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.
Have Questions?
Catalog Number
Quantity
FIS6769001
2 x 1L
Catalog number
FIS6769001
Price (SGD)
-
Quantity:
2 x 1L
Ready-to-use
Thoroughly pH-controlled for completely effective bluing
Specifications
Type
Bluing Reagent
Quantity
2 x 1L