▸
Histology and Cytology
▸
Tissue Fixatives
▸
Formal-Fixx™ 10% Neutral Buffered Formalin
Element Search
Structure Search
Thermo Scientific
Formal-Fixx™ 10% Neutral Buffered Formalin
Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.
Have Questions?
Catalog Number
Quantity
FIS6764254
3.8 L
Catalog number
FIS6764254
Price (SGD)
-
Quantity:
3.8 L
Request bulk or custom format
Specifications
Capacity
Reconstituted to 5 gallon (19 L)
Packaging Type
Ea.
Product Type
Formalin
Quantity
3.8 L
Unit Size
Each
Phosphate-free
Use for storage of all tissue types
Stores specimens without excessive shrinking or hardening
Available premixed, ready-to-use, or in economical concentrate
Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
Pre-filled, leak-resistant cups are available