Check Order Status
Potassium Standard, 100 ppm K, Ricca Chemical
Element Search
Structure Search
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Potassium Standard, 100 ppm K, Ricca Chemical
Have Questions?
Contact Us
Catalog Number
Quantity
FIS6497-16
500 mL
Catalog number
FIS6497-16
Price (SGD)
-
Contact Us
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Standard
Packaging
Natural Poly Bottle
CAS
7447-40-7
CAS Min %
98.0
Concentration or Composition Notes
100ppm K
View more
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link