Check Order Status
Potassium Pyroantimonate TS, Ricca Chemical
Element Search
Structure Search
Potassium Pyroantimonate TS, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS6458-16
500 mL
Natural Poly Bottle
Catalog number
FIS6458-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Pyroantimonate TS
CAS
7732-18-5
CAS Min %
95.0
CAS Max %
99.0
CAS
1310-58-3
View more