Check Order Status
Ammonium Hydroxide, 6.00 N, Ricca Chemical
Element Search
Structure Search
Ammonium Hydroxide, 6.00 N, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS645-16
500 mL
Natural Poly Bottle
Catalog number
FIS645-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ammonium Hydroxide, 6.00N, Ricca Chemical
CAS
1336-21-6
CAS Min %
71.0
CAS Max %
74.0
CAS
7732-18-5
View more