▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Potassium Iodate Volumetric Solution, 0.0500 M (M/20), 0.300 N, Ricca Chemical
Element Search
Structure Search
Potassium Iodate Volumetric Solution, 0.0500 M (M/20), 0.300 N, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS6277-32
1 L
Catalog number
FIS6277-32
Price (SGD)
0.00
Each
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
0.00
Each
Add to cart
Specifications
Chemical Name or Material
POTASSIUM IODATE VOLUMETRIC SOLUTION
CAS
7758-05-6
CAS Min %
1.09
CAS Max %
1.13
CAS
7732-18-5
View more