Acetonitrile 50%, Water 47.5 % and Trifluoroacetic acid 2.5%, For mass spectrometry, Honeywell Riedel-de Haën™
LabReady™ Solvent Blend, for mass spectrometry
Have Questions?
Catalog Number
Quantity
FIS60-048-220
250 mL
Catalog number FIS60-048-220
Price (SGD)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonitrile 50%, Water 47.5% and Trifluoroacetic acid 2.5%
Flash Point<10°C
Shelf Life240 days from date of manufacture
Quantity250 mL
Name NoteFor mass spectrometry
View more
LabReady™ Solvent Blends
Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk