Sodium hydroxide solution, 4M, Honeywell™
Sodium hydroxide solution, 4M, Honeywell™
Sodium hydroxide solution, 4M, Honeywell™
Sodium hydroxide solution, 4M, Honeywell™
Volumetric, 4 M NaOH (4N)
Have Questions?
Catalog NumberQuantity
FIS60-014-571 L
Catalog number FIS60-014-57
Price (SGD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide solution
CAS1310-73-2
Health Hazard 2P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310
Molecular FormulaHNaO
Linear FormulaNaOH
View more
Honeywell´s high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations