Check Order Status
Potassium Chloride Reference Solution A, 111,342 μS/cm at 25°C, 1 M, Ricca Chemical
Structure Search
Potassium Chloride Reference Solution A, 111,342 μS/cm at 25°C, 1 M, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS5893-32
1 L
Catalog number
FIS5893-32
Price (SGD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Chloride Reference Solution A
Packaging
Natural Poly Bottle
Methods
ASTM D 1125 for Conductivity
pH
Approximately 7
Melting Point
Approximately 0°C
View more