Check Order Status
Potassium Chloride Conductivity Standard, 84 μS/cm at 25°C, Ricca Chemical
Element Search
Structure Search
Potassium Chloride Conductivity Standard, 84 μS/cm at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS588684-16
500 mL
Natural Poly Bottle
Catalog number
FIS588684-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Chloride Conductivity Standard
Name Note
84μS/cm (84μmho/cm) at 25°C
Packaging
Natural Poly Bottle
CAS
7447-40-7
CAS Min %
98.0
View more