Check Order Status
Potassium Chloride Conductivity Standard, 12.9 μS/cm at 25°C, Ricca Chemical
Element Search
Structure Search
Potassium Chloride Conductivity Standard, 12.9 μS/cm at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS588513-32
1 L
Catalog number
FIS588513-32
Price (SGD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material
Conductivity Standard
Packaging
Natural Poly Bottle
pH
Approximately 7
Boiling Point
100°C
Solution Type
Conductivity Standard
View more