Check Order Status
Phosphoric Acid, 10% (v/v), Ricca Chemical
Element Search
Structure Search
Phosphoric Acid, 10% (v/v), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS5850-32
1 L
Natural Poly Bottle
Catalog number
FIS5850-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Phosphoric Acid
CAS
7732-18-5
CAS
7664-38-2
Packaging
Natural Poly Bottle
Solvent or Matrix
Aqueous Solution
View more