Check Order Status
p-Naphtholbenzein Indicator, 1% (w/v) in Titration Solvent, Ricca Chemical
Element Search
Structure Search
p-Naphtholbenzein Indicator, 1% (w/v) in Titration Solvent, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS5225-16
500 mL
Amber Glass Bottle
Catalog number
FIS5225-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS
108-88-3, 67-63-0, 145-50-6, 7732-18-5
Specifications
Chemical Name or Material
p-Naphtholbenzein Indicator
CAS
108-88-3
CAS
67-63-0
CAS
145-50-6
CAS
7732-18-5
View more