n-Paraffin Mix C5, C6, C7 & C8 Analytical Standard, Sold by MilliporeSigma™ Supelco™
n-Paraffin Mix C5, C6, C7 & C8 Analytical Standard, Sold by MilliporeSigma™ Supelco™
Fisherbrand
n-Paraffin Mix C5, C6, C7 & C8 Analytical Standard, Sold by MilliporeSigma™ Supelco™
May be used for determining column resolution, retention times, and response factors.
Have Questions?
Catalog NumberQuantity
FIS47-100500 mg
Catalog number FIS47-100
Price (SGD)
-
Quantity:
500 mg
Request bulk or custom format
Specifications
Health Hazard 1H224 - H304 - H315 - H336 - H361f - H373 - H410
Recommended StorageRoom Temperature
Flash Point-17.5°C (Closed Cup)
UN NumberUN3295 - Class 3 - PG 1 - EHS - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
Quantity500 mg
View more
ASTM is a registered trademark of American Society for Testing and Materials