Shandon™ Disposable Base Molds, 0.95 x 0.95 x 0.2 in. (24 x 24 x 5mm)
Shandon™ Disposable Base Molds, 0.95 x 0.95 x 0.2 in. (24 x 24 x 5mm)
Actual product may vary
Thermo Scientific
Shandon™ Disposable Base Molds, 0.95 x 0.95 x 0.2 in. (24 x 24 x 5mm)
Promote rapid paraffin hardening with Epredia™ Shandon™ Disposable Base Molds.
Have Questions?
Catalog NumberLength (Metric)Width (Metric)
FIS41-74115 mm15 mm
Catalog number FIS41-741
Price (SGD)
-
Length (Metric):
15 mm
Width (Metric):
15 mm
  • Constructed of rigid transparent plastic
Specifications
Product TypeBase Mold
MaterialPlastic
Length (Metric)15 mm
Width (Metric)15 mm
Length (English)0.59 in.
Width (English)0.59 in.
Depth (Metric)5 mm
Depth (English)0.2 in.