KleenGuard™ V60 Nemesis™ Safety Eyewear
KleenGuard™ V60 Nemesis™ Safety Eyewear
Actual product may vary
KleenGuard™ V60 Nemesis™ Safety Eyewear
KleenGuard Nemesis Safety Eyewear with Rx Safety Inserts are a convenient way to get prescription eyewear into your workplace. They feature the Nemesis Glasses’ sporty good looks and durable, lightweight construction paired with prescription lenses.
Have Questions?
Catalog NumberFrame ColorFrame TypeLens Tint
FIS3850-4BlackBlack frame with black tipsAmber
Catalog number FIS3850-4
Price (SGD)
-
Frame Color:
Black
Frame Type:
Black frame with black tips
Lens Tint:
Amber
  • Fill prescription at a local vision center and fit the inserts into the safety glasses
  • Tend to be more lightweight and comfortable than options like side shields and over-the-glasses safety glasses
  • Polycarbonate lenses help protect the prescription insert from damage and provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Wraparound frame has a base curve of 6.5 that extends the range of peripheral protection, as well as adjustable ratchet-action temples for with five positions for a customized fit and lens angle
  • Corrective lenses are hard coated for scratch resistance and longer life
  • Each pair comes with a neck cord for securing
  • Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
  • Stylish, comfortable and functional for construction, manufacturing, and a host of other industries
Specifications
Lens CoatingAnti-fog
Certifications/ComplianceMeets ANSI Z87.1+ standards for high impact (38504-00 meets ANSI Z87.1+2010 and 38504-01 meets ANSI Z87.1+2015)
Lens TintAmber
Product TypeSafety Eyewear
Frame ColorBlack
IncludesRx insert
Lens MaterialPolycarbonate
Frame TypeBlack frame with black tips
Unit SizeEA