Hydrochloric Acid, 0.100 N (N/10) in Isopropyl Alcohol, Ricca Chemical
Hydrochloric Acid, 0.100 N (N/10) in Isopropyl Alcohol, Ricca Chemical
Hydrochloric Acid, 0.100 N (N/10) in Isopropyl Alcohol, Ricca Chemical
Hydrochloric Acid, 0.100 N (N/10) in Isopropyl Alcohol, Ricca Chemical
Hydrochloric Acid, 0.100 N (N/10) in Isopropyl Alcohol, Ricca Chemical
Hydrochloric Acid, 0.100 N (N/10) in Isopropyl Alcohol, Ricca Chemical
Have Questions?
Catalog NumberQuantityPackaging
FIS3800-321 LNatural Poly Bottle
Catalog number FIS3800-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric Acid
ColorColorless
Name NoteIn Isopropanol
CAS67-63-0
Concentration0.100Normal
View more