Check Order Status
Hydrochloric Acid, 10% (v/v), Ricca Chemical
Element Search
Structure Search
Hydrochloric Acid, 10% (v/v), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS3570-32
1 L
Natural Poly Bottle
Catalog number
FIS3570-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Hydrochloric Acid
Color
Colorless
CAS
7732-18-5
CAS Min %
88.0
Concentration
10% (v/v)
View more