▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Glycerin (Glycerol), 50% (v/v), Ricca Chemical
Element Search
Structure Search
Glycerin (Glycerol), 50% (v/v), Ricca Chemical
Have Questions?
Change view
Quantity:
1 L
500 mL
Packaging:
Natural Poly Bottle
Catalog number
FIS329032
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Glycerin
Color
Colorless
Name Note
Glycerol
CAS
56-81-5
CAS Min %
55.0
View more
Glycerin (Glycerol), 50% (v/v) Aqueous Solution