Check Order Status
Fluoride Standard, 1 ppm F
-
(Premixed with TISAB/Color Coded Green), Ricca Chemical
Element Search
Structure Search
Fluoride Standard, 1 ppm F
-
(Premixed with TISAB/Color Coded Green), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS3162-16
500 mL
Natural Poly Bottle
Catalog number
FIS3162-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Fluoride Standard
Packaging
Natural Poly Bottle
CAS
7681-49-4
CAS Min %
93.0
CAS Max %
97.0
View more