▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Ferric Chloride CS, USP/EP Yellow Primary Solution, Ricca Chemical
Element Search
Structure Search
Ferric Chloride CS, USP/EP Yellow Primary Solution, Ricca Chemical
1 mL = 45.0 mg FeCl
3
.
6H
2
O
Have Questions?
Catalog Number
Quantity
Packaging
FIS3110-32
1 L
Amber Glass Bottle
Catalog number
FIS3110-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ferric Chloride
Color
Amber
Name Note
Yellow Primary Solution, Colorimetric Solution
CAS
10025-77-1
CAS
7647-01-0
View more